Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978235.1 | XP_027834036.1 | 2 - 3 | 2271 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |