Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A3GALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093608.2 | XP_014949094.2 | 4 - 5 | 1982 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |