Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012162511.3 | XP_012017901.2 | 2 - 3 | 450 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |