Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012162511.3 | XP_012017901.2 | 3 - 4 | 566 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |