Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960210.1 | XP_027816011.1 | 5 - 6 | 422 | 98 | |
XM_027960214.1 | XP_027816015.1 | 5 - 6 | 422 | 98 |