Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008041.4 | XP_004008090.1 | 1 - 2 | 153 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |