Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104605.2 | XP_014960091.2 | 2 - 3 | 411 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |