Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011146.4 | XP_004011195.2 | 6 - 7 | 765 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |