Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961927.1 | XP_027817728.1 | 2 | 298 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |