Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969379.1 | XP_027825180.1 | 2 | 829 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |