Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095581.2 | XP_014951067.2 | 1 | 95 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |