Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979998.1 | XP_027835799.1 | 2 | 742 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |