Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HESX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713403.3 | XP_002713449.1 | 2 - 3 | 685 | 81 | |
GenBank mRNA | AF102132.1 | - | 2 - 3 | 457 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |