Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136522.1 | NP_001129994.1 | 21 - 22 | 3243 | 66 | |
GenBank mRNA | AF199511.1 | - | 7 - 8 | 681 | 66 | |
X15539.1 | - | 3243 | 66 | |||
X55763.1 | - | 3581 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons |