Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719905.3 | XP_002719951.1 | 5 - 6 | 3425 | 87 | |
GenBank mRNA | AF493548.1 | - | 3 - 4 | 400 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons |