Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716536.3 | XP_002716582.1 | 6 - 7 | 976 | 86 | |
GenBank mRNA | DQ122755.1 | - | 3 - 4 | 297 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |