Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082026.1 | NP_001075495.1 | 2 - 3 | 360 | 67 | |
XM_008256716.2 | XP_008254938.1 | 3 - 4 | 696 | 67 | ||
XM_008256717.2 | XP_008254939.1 | 3 - 4 | 463 | 67 | ||
XM_008256718.2 | XP_008254940.1 | 3 - 4 | 689 | 67 | ||
XM_017341617.1 | XP_017197106.1 | 3 - 4 | 444 | 67 | ||
GenBank mRNA | AF022961.1 | - | 2 - 3 | 360 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Oryctolagus cuniculus Probe spans exons |