Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082044.1 | NP_001075513.1 | 24 - 25 | 3046 | 74 | |
XM_008259331.2 | XP_008257553.1 | 25 - 26 | 3060 | 74 | ||
XM_008259332.2 | XP_008257554.1 | 25 - 26 | 3059 | 74 | ||
XM_008259335.2 | XP_008257557.1 | 24 - 25 | 3020 | 74 | ||
XM_008259336.2 | XP_008257558.1 | 24 - 25 | 3021 | 74 | ||
XM_017343242.1 | XP_017198731.1 | 25 - 26 | 3062 | 74 | ||
GenBank mRNA | AF087468.1 | - | 24 - 25 | 3046 | 74 | |
AF417510.1 | - | 22 - 23 | 2773 | 74 | ||
AF417511.1 | - | 22 - 23 | 2773 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |