Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082044.1 | NP_001075513.1 | 6 - 7 | 844 | 79 | |
XM_008259331.2 | XP_008257553.1 | 7 - 8 | 858 | 79 | ||
XM_008259332.2 | XP_008257554.1 | 7 - 8 | 857 | 79 | ||
XM_008259335.2 | XP_008257557.1 | 7 - 8 | 857 | 79 | ||
XM_008259336.2 | XP_008257558.1 | 7 - 8 | 858 | 79 | ||
XM_017343242.1 | XP_017198731.1 | 7 - 8 | 860 | 79 | ||
GenBank mRNA | AF087468.1 | - | 6 - 7 | 844 | 79 | |
AF417510.1 | - | 4 - 5 | 571 | 79 | ||
AF417511.1 | - | 4 - 5 | 571 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |