Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082068.1 | NP_001075537.1 | 10 - 11 | 2151 | 86 | |
XM_008272337.2 | XP_008270559.1 | 14 - 15 | 2828 | 86 | ||
XM_008272338.2 | XP_008270560.1 | 11 - 12 | 2500 | 86 | ||
XM_008272339.2 | XP_008270561.1 | 11 - 12 | 2392 | 86 | ||
GenBank mRNA | AF195523.1 | - | 10 - 11 | 2151 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |