Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082110.1 | NP_001075579.1 | 3 - 4 | 221 | 59 | |
GenBank mRNA | AF372520.1 | - | 3 - 4 | 221 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |