Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082152.1 | NP_001075621.1 | 9 | 2525 | 157 | |
XM_008263958.2 | XP_008262180.1 | 10 | 3018 | 157 | ||
XM_008263959.2 | XP_008262181.1 | 9 | 2451 | 157 | ||
GenBank mRNA | AY212510.1 | - | 9 | 2525 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Oryctolagus cuniculus Both primers and probe map within a single exon |