Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082159.1 | NP_001075628.1 | 21 - 22 | 2830 | 82 | |
XM_008261791.2 | XP_008260013.1 | 22 - 23 | 2893 | 82 | ||
XM_017344296.1 | XP_017199785.1 | 4 - 5 | 478 | 82 | ||
GenBank mRNA | AY360144.3 | - | 21 - 22 | 2830 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons |