Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082159.1 | NP_001075628.1 | 4 - 5 | 425 | 117 | |
XM_008261791.2 | XP_008260013.1 | 5 - 6 | 488 | 117 | ||
GenBank mRNA | AY360144.3 | - | 4 - 5 | 425 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |