Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082207.1 | NP_001075676.1 | 3 - 4 | 543 | 88 | |
XM_017342615.1 | XP_017198104.1 | 3 - 4 | 577 | 88 | ||
GenBank mRNA | D49841.1 | - | 3 - 4 | 543 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |