Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082238.1 | NP_001075707.1 | 5 - 6 | 607 | 58 | |
XM_008257929.2 | XP_008256151.1 | 5 - 6 | 696 | 58 | ||
XM_008257930.2 | XP_008256152.1 | 5 - 6 | 678 | 58 | ||
XM_017342433.1 | XP_017197922.1 | 5 - 6 | 723 | 58 | ||
XM_017342435.1 | XP_017197924.1 | 5 - 6 | 701 | 58 | ||
GenBank mRNA | K02300.1 | - | 5 - 6 | 607 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |