Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082315.1 | NP_001075784.1 | 10 - 11 | 1288 | 75 | |
XM_008248341.2 | XP_008246563.1 | 10 - 11 | 1350 | 75 | ||
XM_008253164.2 | XP_008251386.1 | 9 - 10 | 1250 | 75 | ||
GenBank mRNA | M95815.1 | - | 10 - 11 | 1288 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |