Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082331.1 | NP_001075800.1 | 2 - 3 | 991 | 76 | |
XM_017348407.1 | XP_017203896.1 | 2 - 3 | 3855 | 76 | ||
GenBank mRNA | S82448.1 | - | 2 - 3 | 991 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Oryctolagus cuniculus Probe spans exons |