Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082331.1 | NP_001075800.1 | 4 - 5 | 1350 | 65 | |
XM_017348407.1 | XP_017203896.1 | 4 - 5 | 4127 | 65 | ||
GenBank mRNA | S82448.1 | - | 4 - 5 | 1350 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |