Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082339.1 | NP_001075808.1 | 3 - 4 | 665 | 67 | |
GenBank mRNA | U09823.1 | - | 3 - 4 | 665 | 67 | |
X62245.1 | - | 635 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |