Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Oc03398120_g1
See other SLMAP GE Assays ›
Gene Symbol:
SLMAP
Gene Name:
sarcolemma associated protein
Gene Aliases:
SLAP
Chromosome Location:
Chr.9: 24582901 - 24751719 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID
Oc03398120_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SLMAP
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001082348.2
NP_001075817.2
13 - 14
1342
83
XM_008260454.2
XP_008258676.1
16 - 17
2680
83
XM_008260455.2
XP_008258677.1
16 - 17
2674
83
XM_008260456.2
XP_008258678.1
16 - 17
2680
83
XM_008260457.2
XP_008258679.1
15 - 16
2629
83
XM_008260458.2
XP_008258680.1
15 - 16
2629
83
XM_008260459.2
XP_008258681.1
15 - 16
2623
83
XM_008260460.2
XP_008258682.1
15 - 16
2617
83
XM_008260461.2
XP_008258683.1
15 - 16
2611
83
XM_008260462.2
XP_008258684.1
15 - 16
2611
83
XM_008260463.2
XP_008258685.1
14 - 15
2578
83
XM_008260464.2
XP_008258686.1
14 - 15
2566
83
XM_008260465.2
XP_008258687.1
14 - 15
2566
83
XM_008260466.2
XP_008258688.1
16 - 17
2682
83
XM_008260467.2
XP_008258689.1
13 - 14
2514
83
XM_008260468.2
XP_008258690.1
13 - 14
2508
83
XM_008260470.2
XP_008258692.1
14 - 15
2580
83
XM_008260471.2
XP_008258693.1
14 - 15
2573
83
XM_008260472.2
XP_008258694.1
13 - 14
2516
83
XM_008260473.2
XP_008258695.1
13 - 14
2510
83
XM_017343638.1
XP_017199127.1
14 - 15
2572
83
XM_017343639.1
XP_017199128.1
16 - 17
2685
83
XM_017343640.1
XP_017199129.1
2 - 3
221
83
XM_017343641.1
XP_017199130.1
2 - 3
242
83
GenBank mRNA
AF304449.1
-
2 - 3
183
83
U21155.1
-
2 - 3
183
83
U21156.1
-
3 - 4
255
83
U21157.1
-
13 - 14
1703
83
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017343640.1, XM_017343641.1 transcripts.
Set Membership:
Amplicon length between 71 and 85
Detect Genomic DNA
Oryctolagus cuniculus
Probe spans exons
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0