Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082381.1 | NP_001075850.1 | 1718 | 111 | ||
GenBank mRNA | U82163.1 | - | 1718 | 111 |
Gene Symbol: LOC103345956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017339189.1 | XP_017194678.1 | 861 | 111 |
Gene Symbol: LOC103345957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017339169.1 | XP_017194658.1 | 954 | 111 |
Gene Symbol: LOC103345959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008251337.2 | XP_008249559.1 | 1173 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus |