Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082408.1 | NP_001075877.1 | 2 - 3 | 378 | 79 | |
XM_008271895.2 | XP_008270117.1 | 2 - 3 | 393 | 79 | ||
GenBank mRNA | X98303.1 | - | 378 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |