Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082633.1 | NP_001076102.1 | 1 - 2 | 121 | 107 | |
GenBank mRNA | AB016237.1 | - | 1 - 2 | 121 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Oryctolagus cuniculus Probe spans exons |