Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082637.1 | NP_001076106.1 | 12 - 13 | 1681 | 67 | |
GenBank mRNA | AF080067.1 | - | 12 - 13 | 1681 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons |