Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082703.2 | NP_001076172.1 | 12 - 13 | 1648 | 68 | |
XM_008257419.1 | XP_008255641.1 | 13 - 14 | 68 | |||
XM_017342181.1 | XP_017197670.1 | 8 - 9 | 68 | |||
GenBank mRNA | U55053.1 | - | 12 - 13 | 1649 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |