Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082728.1 | NP_001076197.1 | 11 - 12 | 1518 | 135 | |
GenBank mRNA | AJ132108.1 | - | 11 - 12 | 1518 | 135 | |
AY521461.1 | - | 10 - 11 | 1501 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |