Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082757.1 | NP_001076226.1 | 28 - 29 | 3905 | 145 | |
GenBank mRNA | M59743.1 | - | 28 - 29 | 3892 | 145 | |
U50465.1 | - | 28 - 29 | 3905 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |