Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082649.1 | NP_001076118.1 | 13 - 14 | 1817 | 69 | |
GenBank mRNA | AF028241.1 | - | 13 - 14 | 1817 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |