Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082661.1 | NP_001076130.1 | 5 - 6 | 849 | 102 | |
GenBank mRNA | AY561276.1 | - | 5 - 6 | 849 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |