Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082724.1 | NP_001076193.1 | 1 - 2 | 111 | 83 | |
XM_008262720.2 | XP_008260942.1 | 2 - 3 | 205 | 83 | ||
GenBank mRNA | AF318299.1 | - | 1 - 2 | 111 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |