Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082750.1 | NP_001076219.1 | 11 - 12 | 2218 | 77 | |
XM_008259342.2 | XP_008257564.1 | 11 - 12 | 2257 | 77 | ||
XM_017343246.1 | XP_017198735.1 | 11 - 12 | 2170 | 77 | ||
GenBank mRNA | U09490.1 | - | 11 - 12 | 2218 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |