Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099957.1 | NP_001093427.1 | 2 - 3 | 575 | 83 | |
GenBank mRNA | EF062856.1 | - | 2 - 3 | 575 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |