Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099957.1 | NP_001093427.1 | 3 - 4 | 702 | 55 | |
GenBank mRNA | AB182322.1 | - | 2 - 3 | 191 | 55 | |
EF062856.1 | - | 3 - 4 | 702 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |