Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFIP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099965.1 | NP_001093435.1 | 1998 | 83 | ||
XM_008274606.2 | XP_008272828.1 | 2103 | 83 | |||
XM_008274607.2 | XP_008272829.1 | 2321 | 83 | |||
XM_017350649.1 | XP_017206138.1 | 1 | 1758 | 83 | ||
GenBank mRNA | EF443269.1 | - | 1998 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Both primers and probe map within a single exon |