Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082324.1 | NP_001075793.1 | 2 | 1843 | 86 | |
XM_008266172.2 | XP_008264394.1 | 3 | 2009 | 86 | ||
XM_008266173.2 | XP_008264395.1 | 3 | 1946 | 86 | ||
XM_008266174.2 | XP_008264396.1 | 2 | 1779 | 86 | ||
GenBank mRNA | S59041.1 | - | 2 | 1843 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |