Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082332.2 | NP_001075801.1 | 49 | 78 | ||
GenBank mRNA | U01117.1 | - | 49 | 78 |
Gene Symbol: FOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274515.1 | XP_008272737.1 | 719 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus Inventoried |