Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082399.1 | NP_001075868.1 | 1 | 47 | 63 | |
GenBank mRNA | X74728.1 | - | 47 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Oryctolagus cuniculus Both primers and probe map within a single exon |