Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082652.1 | NP_001076121.1 | 1 | 949 | 73 | |
GenBank mRNA | AF209723.1 | - | 1 | 949 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |