Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017341428.1 | XP_017196917.1 | 88 | |||
GenBank mRNA | M10412.1 | - | 281 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus |